Aluminum phosphate ≥99.99%, Puratronic®
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Aluminium phosphate
10628-06
10628-14
CAAA10628-06EA
151.84
CAD
CAAA10628-06
CAAA10628-14
Aluminum phosphate ≥99.99%, Puratronic®
Aluminum phosphate
Purity: 99.99%
CAS No.: 7784-30-7
Molecular Formula: AlO4P
Formula Weight: 121.95
Physical Form: Powder
Density: 2.566
Appearance: White
Melting Point:
Formula:
AlPO₄ Melting Pt: >1500 °C Density: 2.566 Storage Temperature: Ambient |
MDL Number:
MFCD00003430 CAS Number: 7784-30-7 EINECS: 232-056-9 UN: 3260 ADR: 8,III Merck Index: 14,358 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...