Tetraammineplatinum(II) chloride monohydrate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
10544-03
10544-06
CAAA10544-03EA
298.62
CAD
CAAA10544-03
CAAA10544-06
Tetraammineplatinum(II) chloride monohydrate ≥98%
Tetraammineplatinum(II) chloride monohydrate
CAS No.: 13933-33-0
Molecular Formula: Pt(NH3)4Cl2.H2O
Formula Weight: 352.13 (334.11anhy)
Physical Form: Crystalline
Density: 2.74
Sensitivity: Hygroscopic
Appearance: White to cream
Melting Point: 250° dec.
MDL No.: MFCD00149947
Formula:
Cl₂H₁₄N₄OPt Melting Pt: 250 °C (dec.) Density: 2.74 Storage Temperature: Ambient |
MDL Number:
MFCD00149947 CAS Number: 13933-33-0 EINECS: 237-706-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...