Potassium tetracyanoplatinate(II) trihydrate (≥44,9% Pt) ≥99.9%

Supplier: Thermo Scientific Chemicals
Danger

10536-03 10536-06
CAAA10536-03EA 281.24 CAD
CAAA10536-03 CAAA10536-06
Potassium tetracyanoplatinate(II) trihydrate (≥44,9% Pt) ≥99.9%
Potassium tetracyanoplatinate (II) trihydrate

Potassium tetracyanoplatinate(II) trihydrate is used in the preparation other salts like Krogmann's salt, that are highly conducting, one dimensional materials.


Soluble in hot water

Formula: C₄K₂N₄Pt·3H₂O
MW: 431.39
Storage Temperature: Ambient
MDL Number: MFCD00149921
CAS Number: 38245-27-1
EINECS: 209-236-0
UN: 1588
ADR: 6.1 III
Merck Index: 14,7685

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR