Dodecacarbonyltriruthenium 99%

Supplier: Thermo Scientific Chemicals
Danger

10507-03 10507-06
CAAA10507-03EA 208.56 CAD
CAAA10507-03 CAAA10507-06
Dodecacarbonyltriruthenium 99%
Dodecacarbonyltriruthenium

Application: Carbonylation


Sparingly soluble in hydrocarbons (e.g. hexane, cyclohexane, benzene) and acetone. Solutions undergo some decomposition on strong heating


Carbonylation

Formula: C₁₂O₁₂Ru₃
Melting Pt: ∼ 150 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011209
CAS Number: 15243-33-1
EINECS: 239-287-4
UN: 3466
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR