Dichloro(ethylenediamine)palladium(II) (44,8% Pd)
Supplier: Thermo Scientific Chemicals
|
Warning
|
10496-03
10496-06
CAAA10496-03EA
230.68
CAD
CAAA10496-03
CAAA10496-06
Dichloro(ethylenediamine)palladium(II) (44,8% Pd)
Dichloro(ethylenediamine)palladium(II)
CAS No.: 15020-99-2
Molecular Formula: PdCl2(H2NCH2CH2NH2)
Formula Weight: 237.41
Physical Form: Crystalline
Sensitivity: Hygroscopic
MDL No.: MFCD00044991
Formula:
PdCl₂(H₂NCH₂CH₂NH₂) Storage Temperature: Ambient |
MDL Number:
MFCD00044991 CAS Number: 15020-99-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...