Dichloro(1,5-cyclooctadiene)palladium(II) (36,7% Pd)

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,5-Cyclooctadienepalladium(II) Dichloride

10493-03 10493-06
CAAA10493-03EA 232.26 CAD
CAAA10493-03 CAAA10493-06
Dichloro(1,5-cyclooctadiene)palladium(II) (36,7% Pd)
Dichloro(1,5-cyclooctadiene)palladium(II)

Soluble in dichloromethane


CAS No.: 12107-56-1 Molecular Formula: C8H12Cl2Pd Formula Weight: 285.49 Physical Form: Crystalline Sensitivity: Hygroscopic Appearance: Yellow Solubility: Soluble in dichloromethane MDL No.: MFCD00012412

Formula: C₈H₁₂Cl₂Pd
MW: 285.50 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00012412
CAS Number: 12107-56-1
EINECS: 235-161-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR