trans-Dichlorobis(diethylsulfide)platinum(II) (43,7% Pt)

Supplier: Thermo Scientific Chemicals
Warning

10484-03 10484-06
CAAA10484-03EA 180.12 CAD
CAAA10484-03 CAAA10484-06
trans-Dichlorobis(diethylsulfide)platinum(II) (43,7% Pt)
trans-Dichlorobis(diethylsulfide)platinum(II)

Applications: Hydrosilation
Soluble in acetone, alcohol and benzene


CAS No.: 15337-84-5 Molecular Formula: Pt[(C2H5)2S]2Cl2 Formula Weight: 446.37 Physical Form: Crystalline Solubility: Soluble in acetone, alcohol and benzene MDL No.: MFCD00050943


Applications: Hydrosilation

Formula: Pt[(C₂H₅)₂S]₂Cl₂
Storage Temperature: Ambient
MDL Number: MFCD00050943
CAS Number: 15337-84-5
EINECS: 239-373-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR