cis-Dichlorobis(diethylsulfide)platinum(II) (43,7% Pt)

Supplier: Thermo Scientific Chemicals
Warning

10482-02 10482-03
CAAA10482-02EA 120.24 CAD
CAAA10482-02 CAAA10482-03
cis-Dichlorobis(diethylsulfide)platinum(II) (43,7% Pt)
cis-Dichlorobis(diethylsulfide)platinum(II)

Soluble in acetone, alcohol and benzene


Applications: Hydrogenation


Applications: Hydrogenation

Formula: Pt[(C₂H₅)₂S]₂Cl₂
Storage Temperature: Ambient
MDL Number: MFCD00050943
CAS Number: 15442-57-6
EINECS: 239-454-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR