Diethyl (trichloromethyl)phosphonate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Diethyltrichloromethylphosphonate, (Trichloromethyl)phosphonic Acid Diethyl Ester
10306-04
10306-09
CAAA10306-04EA
80.74
CAD
CAAA10306-04
CAAA10306-09
Diethyl (trichloromethyl)phosphonate ≥98%
Diethyl (trichloromethyl)phosphonate
Purity: 98%
CAS No.: 866-23-9
Molecular Formula: (C2H5O)2P(O)(CCl3)
Formula Weight: 255.48
Physical Form: Liquid
Density: 1.3670
Refractive Index: 1.4620
Boiling Point: 128°/14mm
UN No.: 3278
MDL No.: MFCD00013666
Formula:
C₅H₁₀Cl₃O₃P Boiling Pt: 128 °C (14 mmHg) Density: 1.3670 Storage Temperature: Ambient |
MDL Number:
MFCD00013666 CAS Number: 866-23-9 UN: 3278 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...