Tungsten rhenium ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)
Supplier: Thermo Scientific Chemicals
10073-G2
10073-G6
CAAA10073-G2EA
140.46
CAD
CAAA10073-G2
CAAA10073-G6
Tungsten rhenium ≥99.95% (metals basis), wire, Ø 0.25 mm (0.01 in)
Tungsten rhenium
Purity: 99.95%
Molecular Formula: W:Re; 97:3 wt%
Physical Form: ≈1.9g/2m
MDL No.: MFCD02091751
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...