Allylpalladium(II) chloride dimer (≥ 56,0% Pd)

Supplier: Thermo Scientific Chemicals
Warning

10005-01 10005-03 10005-06
CAAA10005-01EA 95.59 CAD
CAAA10005-01 CAAA10005-03 CAAA10005-06
Allylpalladium(II) chloride dimer (≥ 56,0% Pd)
Allylpalladium(II) chloride dimer
Formula: C₆H₁₀Cl₂Pd₂
Melting Pt: 120 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00044874
CAS Number: 12012-95-2
EINECS: 234-579-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR