Bis(acetonitrile)palladium(II) dichloride (40.5% Pd)

Supplier: Thermo Scientific Chemicals
Danger

10002-03 10002-06
CAAA10002-03EA 213.3 CAD
CAAA10002-03 CAAA10002-06
Bis(acetonitrile)palladium(II) dichloride (40.5% Pd)
Bis(acetonitrile)palladium(II) dichloride

For olefin isomerization, preparation of acetals and hemiacetal esters.
Soluble in acetonitrile, acetone, chloroform.


CAS No.: 14592-56-4 Molecular Formula: PdCl2(CH3CN)2 Formula Weight: 259.41 Physical Form: Powder Odor: Odorless Solubility: Soluble in acetonitrile, acetone, chloroform UN No.: 3439 MDL No.: MFCD00013122


Applications: Olefin isomerization, preparation of acetals and hemiacetal esters

Formula: C₄H₆Cl₂N₂Pd
Storage Temperature: Ambient
MDL Number: MFCD00013122
CAS Number: 14592-56-4
EINECS: 238-637-3
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR