Tweezers, Angled Fine Style 51, Excelta Corp®
Supplier: Excelta
51-S 5C-SA 51-SA-PI 51-SA-PI-ET
63042-510EA 191.75 CAD
63042-510 63042-682 94024-284 89411-808
Tweezers, Angled Fine Style 51, Excelta Corp®
Forceps
The offset oblique tips offer excellent visibility under a scope.
  • Precision points for work under a scope
  • Offset tips offer excellent visibility
  • Offset tips excellent for accessibility in tight places
The '51 Style' series 45° offset oblique angle micro-fine tipped tweezer is ideal for working under a scope. It is also available with our Ergo-Tweeze® soft cushioned 'ET' grips (10¹⁰ ohms/sq surface resistivity), which also protect the user from heat. There is also a 30° offset style available. We offer 2 different qualities. The higher the quality, theMore Product Information