VWR® Contour™ Adjustable Slip Scribes for Base Cabinets, Steel
Supplier: VWR International
CFL-3012-BA CFL-3412-BA CFL-3612-BA CWL-3012-BA CWL-3412-BA CWL-3612-BA
CFL-3012-BAEA 522.79 CAD
CFL-3012-BA CFL-3412-BA CFL-3612-BA CWL-3012-BA CWL-3412-BA CWL-3612-BA
VWR® Contour™ Adjustable Slip Scribes for Base Cabinets, Steel
Furniture Casework
Steel adjustable slip scribes are designed the same as standard scribes with the addition of a slip-on angle at the wall edge to dress-up the edge of the scribe.
  • Baked, chemical-resistant, powder-coat finish
  • Light neutral color
The steel adjustable slip scribe is designed to be cut to width in the field to more easily fit the space between the wall and the end of the furniture assembly.
Order Now
SPECIFICATIONS
Environmentally Preferable