Tweezers, Strong with Serrated Handle, Excelta Corp®
Supplier: Excelta
AC-PI AC-SA AC-SA-PI
63043-218EA 65.51 CAD
63043-218 63043-222 63043-224
Tweezers, Strong with Serrated Handle, Excelta Corp®
Forceps
Serrated handles make these tweezers much easier to handle.
  • Serrated handle for positive control
  • Strong tips that won't bend
  • Ideal for general assembly
The 'Style AC' Series straight medium pointed tweezers are general purpose tweezer used without magnification. The series features robust medium pointed tips for longevity and serrated handles for more positive control. We offer 3 different qualities. The higher the quality, the more precise the tips. They are offered in carbon steel (hard), stainless steel (rust-resistant) or stainless anti-magnetic steel (anti-magnetic).