Five-Star™ Cutters, Medium Head, Excelta
Supplier: Excelta
7140E 7143E 7144E 7146E 7148E 7240E 7241E 7242E 7244E 7245E 7246E 7247E 7248E
63042-782EA 239.34 CAD
63042-782 63042-784 63042-786 63042-788 63042-790 63042-792 63042-794 63042-796 63042-798 63042-834 63042-836 63042-838 63042-842 63042-844 63042-846 63042-848 63042-850
Five-Star™ Cutters, Medium Head, Excelta
Cutters
Five-Star™ 7000 series medium head cutters are precision side cutters designed to make an extremely precise cut on small leads and soft wire.
  • Available either with long foam cushion grips or ESD safe molded grips
  • The dual stainless-steel leaf springs provide smooth action
These are feature with raised bearing surface, Rockwell C 63-65 hardness, and a Delrin washer for smooth operation. The CNC machining allows the joint surfaces to be perfect to allow for long life.