Craig Recrystallization Tube, Microscale, Ace Glass Incorporated
Supplier: Ace Glass
9581-03 9581-05 9581-23 9581-24 9581-28 9581-30
89075-194EA 81.04 CAD
89075-194 89075-196 89075-198 89075-200 89075-202 89075-204
Craig Recrystallization Tube, Microscale, Ace Glass Incorporated
Tubes Recrystallization Tubes
Microscale Craig tube for recrystallizing small quantities of reaction product.
The assembly consists of a glass outer tube, with non-uniform grind to purposely allow leakage, and a PTFE inner plug. The plug eliminates breakage normally experienced with glass plugs. Stated volumes indicate the capacity below the grind. Fabricated from borosilicate glass.
Ordering information: Tube is available complete including glass outer member and PTFE inner plug, or as individual components that must be purchased separately. A glass inner plug is also available in lieu of PTFE.