Cutters, Long Nose Fine Tip, Excelta
Supplier: Excelta
7170EA 7171E 7270E 7270EA 7271E 7149E 7249E 7371E
63042-804EA 353.16 CAD
63042-804 63042-806 63042-808 63042-856 63042-858 63042-864 94024-328 94024-360 76471-626
Cutters, Long Nose Fine Tip, Excelta
Cutters
Offers excellent visibility and accessibility for tight cutting of small items.
  • Reaches into very tight places
  • Ergonomic grips/dual leaf springs
  • Smallest tips available on a cutter
  • Induction hardened for longevity
The Lazer Line® medium fine tip optimum flush cutters are ideal for cutting soft small wire in confined areas. They are made of carbon tool steel with induction hardened cutting blades to RHC 63-65. The CNC machining allows the joint surfaces to be perfect to allow for long life. The grips are soft cushioned foam and have a surfaceMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable