Specific Gravity Set

Supplier: VWR International
PH0112A PH0112B PH0112E PH0112D
470137-628PK 45.5 CAD
470137-628 470137-630 470137-632 470137-634 470137-636
Specific Gravity Set
Educational Materials Physics Educational Materials Mechanics Learning Activities
Teach About Gravity

  • Metals included are: aluminum, brass, copper and steel
  • Each cylinder is 5cm long and 13mm diameter
  • Purchase as a set or individually

Determine the gravity of common metals using this set of specific gravity cylinders. Perfect for specific gravity determinations in the classroom. 

Caution: WARNING: CHOKING HAZARD-Not for children under 3 years.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR