Naphthalene-1,4-dicarbonitrile 98+%
Supplier: Thermo Scientific Chemicals
L20248-06
L20248-14
CAAAAL20248-06EA
90.69
CAD
CAAAAL20248-06
CAAAAL20248-14
CAAAAL20248-22
Naphthalene-1,4-dicarbonitrile 98+%
Naphthalene-1,4-dicarbonitrile
Purity: 98+%
CAS No.: 3029-30-9
Molecular Formula: C12H6N2
Formula Weight: 178.19
Odor: Odorless
Melting Point: 208-212°
Beilstein Reference: 1870226
MDL No.: MFCD03093626
Formula:
C₁₂H₆N₂ MW: 178.19 g/mol Melting Pt: 208…212 °C Storage Temperature: Ambient |
MDL Number:
MFCD03093626 CAS Number: 3029-30-9 EINECS: 221-197-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...