4-Aminopiperidine 95%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L20127-06
L20127-14
CAAAAL20127-06EA
119.61
CAD
CAAAAL20127-06
CAAAAL20127-14
4-Aminopiperidine 95%
4-Aminopiperidine
Purity: 98%
CAS No.: 13035-19-3
Molecular Formula: C5H12N2
Formula Weight: 100.16
Flash Point: 46°(114°F)
Refractive Index: 1.4910
Boiling Point: 65°/18mm
Beilstein Reference: 1105
UN No.: 2733
MDL No.: MFCD02179399
Formula:
C₅H₁₂N₂ Boiling Pt: 65 °C/18 mm Density: 0.954 Flash Pt: 46 °C(114 °F) Storage Temperature: Ambient |
MDL Number:
MFCD02179399 CAS Number: 13035-19-3 UN: 2733 ADR: "3",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...