3-Fluoroisonicotinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L20042-03
L20042-06
CAAAAL20042-03EA
47.24
CAD
CAAAAL20042-03
CAAAAL20042-06
CAAAAL20042-MD
3-Fluoroisonicotinic acid 97%
3-Fluoroisonicotinic acid
Purity: 97%
CAS No.: 393-53-3
Molecular Formula: C6H4FNO2
Formula Weight: 141.10
Melting Point: 251-254° dec.
Beilstein Reference: 383678
Formula:
C₆H₄FNO₂ Melting Pt: 251…254 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD03789251 CAS Number: 393-53-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...