2-Bromoisonicotinic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

L19855-03 L19855-06
CAAAAL19855-03EA 104.91 CAD
CAAAAL19855-03 CAAAAL19855-06
2-Bromoisonicotinic acid 97%
2-Bromoisonicotinic acid

Purity: 97% CAS No.: 66572-56-3 Molecular Formula: C6H4BrNO2 Formula Weight: 202.01 Melting Point: ca 225° dec. Beilstein Reference: 471895

Formula: C₆H₄BrNO₂
Melting Pt: ∼ 225 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01646069
CAS Number: 66572-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR