3-Fluoro-4-methoxyphenylboronic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19818-03
L19818-06
CAAAAL19818-03EA
76.31
CAD
CAAAAL19818-03
CAAAAL19818-06
3-Fluoro-4-methoxyphenylboronic acid 98+%
3-Fluoro-4-methoxyphenylboronic acid
Purity: 98+%
CAS No.: 149507-26-6
Molecular Formula: C7H8BFO3
Formula Weight: 169.95
Melting Point: 237-239°
Beilstein Reference: 7370120
Formula:
C₇H₈BFO₃ Melting Pt: 237…239 °C Storage Temperature: Ambient |
MDL Number:
MFCD00807404 CAS Number: 149507-26-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...