3,5-Difluoro-2-methoxyphenylboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19773-03
L19773-06
L19773-MD
CAAAAL19773-03EA
107.12
CAD
CAAAAL19773-03
CAAAAL19773-06
CAAAAL19773-MD
3,5-Difluoro-2-methoxyphenylboronic acid 97%
3,5-Difluoro-2-methoxyphenylboronic acid
Purity: 97%
CAS No.: 737000-76-9
Molecular Formula: C7H7BF2O3
Formula Weight: 187.94
Melting Point: 112-115°
MDL No.: MFCD03095353
Formula:
C₇H₇BF₂O₃ Melting Pt: 112…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD03095353 CAS Number: 737000-76-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...