4-(Methylamino)pyridine 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19626-03
L19626-06
L19626-14
CAAAAL19626-03EA
77.26
CAD
CAAAAL19626-03
CAAAAL19626-06
CAAAAL19626-14
4-(Methylamino)pyridine 99%
4-(Methylamino)pyridine
Purity: 99%
CAS No.: 1121-58-0
Molecular Formula: C6H8N2
Formula Weight: 108.14
Melting Point: 124-128°
Beilstein Reference: 108213
Formula:
C₆H₈N₂ Melting Pt: 124…128 °C Storage Temperature: Ambient |
MDL Number:
MFCD00152625 CAS Number: 1121-58-0 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...