2-Bromo-6-formylpyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

L19518-03 L19518-06 L19518-14
CAAAAL19518-03EA 71.42 CAD
CAAAAL19518-03 CAAAAL19518-06 CAAAAL19518-14
2-Bromo-6-formylpyridine 97%
2-Bromo-6-formylpyridine

Purity: 97% CAS No.: 34160-40-2 Molecular Formula: C6H4BrNO Formula Weight: 186.01 Melting Point: 79-83° Beilstein Reference: 1524300

Formula: C₆H₄BrNO
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD02683546
CAS Number: 34160-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR