Imidazole-2-carboxylic acid ethyl ester 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Ethyl imidazole-2-carboxylate, Ethyl 1H-imidazole-2-carboxylate
L19405-03
L19405-06
CAAAAL19405-03EA
127.98
CAD
CAAAAL19405-03
CAAAAL19405-06
Imidazole-2-carboxylic acid ethyl ester 97%
Imidazole-2-carboxylic acid ethyl ester
Purity: 97%
CAS No.: 33543-78-1
Molecular Formula: C6H8N2O2
Formula Weight: 140.14
Melting Point: 176-178°
Boiling Point: 48-51°/0.3mm
MDL No.: MFCD03426031
Formula:
C₆H₈N₂O₂ MW: 140.14 g/mol Boiling Pt: 48…51 °C/0.3 mm Melting Pt: 176…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD03426031 CAS Number: 33543-78-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...