3-Fluoro-4-hydroxybenzaldehyde 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L19325-03
L19325-06
CAAAAL19325-03EA
156.1
CAD
CAAAAL19325-03
CAAAAL19325-06
3-Fluoro-4-hydroxybenzaldehyde 98%
3-Fluoro-4-hydroxybenzaldehyde
Purity: 98%
CAS No.: 405-05-0
Molecular Formula: C7H5FO2
Formula Weight: 140.11
Melting Point: 121-124°
Beilstein Reference: 3234259
Formula:
C₇H₅FO₂ MW: 140.11 g/mol Melting Pt: 121…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016626 CAS Number: 405-05-0 |
Learn more
![](/m-ca.vwr.com/en_CA/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...