(6-Chloro-3-pyridinyl)methanol 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
6-Chloropyridine-3-methanol, 2-Chloro-5-hydroxymethylpyridine, 6-Chloro-3-(hydroxymethyl)pyridine
L19285-06
L19285-14
CAAAAL19285-06EA
138.25
CAD
CAAAAL19285-06
CAAAAL19285-14
(6-Chloro-3-pyridinyl)methanol 98%
(6-Chloro-3-pyridinyl)methanol
Purity: 98%
CAS No.: 21543-49-7
Molecular Formula: C6H6ClNO
Formula Weight: 143.57
Melting Point: 46-49°
Boiling Point: 120°/0.4mm
MDL No.: MFCD01075226
Formula:
C₆H₆ClNO MW: 143.57 g/mol Boiling Pt: 120 °C/0.4 mm Melting Pt: 46…49 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075226 CAS Number: 21543-49-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...