5-(Aminomethyl)-2-chloropyridine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19283-03
L19283-06
L19283-14
CAAAAL19283-03EA
80.9
CAD
CAAAAL19283-03
CAAAAL19283-06
CAAAAL19283-14
5-(Aminomethyl)-2-chloropyridine 97%
5-(Aminomethyl)-2-chloropyridine
Purity: 97%
CAS No.: 97004-04-1
Molecular Formula: C6H7ClN2
Formula Weight: 142.59
Melting Point: 27-32°
Boiling Point: 101-102°/1mm
UN No.: 3259
MDL No.: MFCD00673153
Formula:
C₆H₇ClN₂ Boiling Pt: 101…102 °C/1 mm Melting Pt: 27…32 °C Storage Temperature: Ambient |
MDL Number:
MFCD00673153 CAS Number: 97004-04-1 UN: 3259 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...