Ethyl-4-(bromomethyl)benzoate 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L19219-06
L19219-14
L19219-22
CAAAAL19219-06EA
67.15
CAD
CAAAAL19219-06
CAAAAL19219-14
CAAAAL19219-22
Ethyl-4-(bromomethyl)benzoate 96%
Ethyl 4-(bromomethyl)benzoate
Purity: 96%
CAS No.: 26496-94-6
Molecular Formula: C10H11BrO2
Formula Weight: 243.10
Storage Temperature: 2°C to 8°C
Refractive Index: 1.5494
Melting Point: 28-33°
Boiling Point: 145°/5mm
Beilstein Reference: 1869569
UN No.: 1759
MDL No.: MFCD03791307
Formula:
C₁₀H₁₁BrO₂ Boiling Pt: 145 °C/5 mm Melting Pt: 28…33 °C Storage Temperature: Ambient |
MDL Number:
MFCD03791307 CAS Number: 26496-94-6 UN: 1759 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...