2-Methylindol-3-ylacetic acid 98+%
Supplier: Thermo Scientific Chemicals
L19071-03
L19071-06
CAAAAL19071-03EA
47.56
CAD
CAAAAL19071-03
CAAAAL19071-06
CAAAAL19071-14
2-Methylindol-3-ylacetic acid 98+%
2-Methylindol-3-ylacetic acid
Purity: 98+%
CAS No.: 1912-43-2
Molecular Formula: C11H11NO2
Formula Weight: 189.22
Melting Point: 198-204° dec.
Beilstein Reference: 152701
Formula:
C₁₁H₁₁NO₂ Melting Pt: 198…204 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00075006 CAS Number: 1912-43-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...