Diethyl octylphosphonate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Diethyl 1-octylphosphonate

L18588-06 L18588-14 L18588-22
CAAAAL18588-06EA 102.23 CAD
CAAAAL18588-06 CAAAAL18588-14 CAAAAL18588-22
Diethyl octylphosphonate 98%
Diethyl octylphosphonate

Purity: 98+% CAS No.: 1068-07-1 Molecular Formula: C12H27O3P Formula Weight: 250.32 Refractive Index: 1.4340 Boiling Point: 88-89°/0.1mm MDL No.: MFCD00015659

Formula: C₁₂H₂₇O₃P
Boiling Pt: 88…89 °C/0.1 mm
Density: 0.953
Storage Temperature: Ambient
MDL Number: MFCD00015659
CAS Number: 1068-07-1
EINECS: 213-941-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR