2-(Methoxycarbonyl)phenylboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methoxycarbonylphenylboronic acid, 2-(Methoxycarbonyl)benzeneboronic acid

L17958-03 L17958-06
CAAAAL17958-03EA 57.83 CAD
CAAAAL17958-03 CAAAAL17958-06 CAAAAL17958-MD
2-(Methoxycarbonyl)phenylboronic acid 97%
2-(Methoxycarbonyl)phenylboronic acid

Purity: 97% CAS No.: 374538-03-1 Molecular Formula: C8H9BO4 Formula Weight: 179.97 Melting Point: 68-70° Beilstein Reference: 8313459

Formula: C₈H₉BO₄
MW: 179.97 g/mol
Melting Pt: 68…70 °C
Storage Temperature: Ambient
MDL Number: MFCD02179452
CAS Number: 374538-03-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR