1-Boc-3-hydroxypiperidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17600-03
L17600-06
CAAAAL17600-03EA
71.1
CAD
CAAAAL17600-03
CAAAAL17600-06
1-Boc-3-hydroxypiperidine 97%
1-Boc-3-hydroxypiperidine
Purity: 97%
CAS No.: 85275-45-2
Molecular Formula: C10H19NO3
Formula Weight: 201.27
Melting Point: 66-70°
Beilstein Reference: 4180859
Formula:
C₁₀H₁₉NO₃ Melting Pt: 66…70 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093938 CAS Number: 85275-45-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...