5-Iodo-1,2,3-trimethoxybenzene 98+%
Supplier: Thermo Scientific Chemicals
L17414-03
L17414-06
CAAAAL17414-03EA
127.35
CAD
CAAAAL17414-03
CAAAAL17414-06
5-Iodo-1,2,3-trimethoxybenzene 98+%
5-Iodo-1,2,3-trimethoxybenzene
Purity: 98+%
CAS No.: 25245-29-8
Molecular Formula: C9H11IO3
Formula Weight: 294.09
Melting Point: 85-87°
Beilstein Reference: 1529690
Formula:
C₉H₁₁IO₃ Melting Pt: 85…87 °C Storage Temperature: Ambient |
MDL Number:
MFCD01318153 CAS Number: 25245-29-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...