(3,3,3-Trifluoropropyl)dimethoxymethylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Dimethoxy(methyl)(3,3,3-trifluoropropyl)silane, 1,1,1-Trifluoro-3-[dimethoxy(methyl)silyl]propane
L17292-06
L17292-14
CAAAAL17292-06EA
96.85
CAD
CAAAAL17292-06
CAAAAL17292-14
CAAAAL17292-22
(3,3,3-Trifluoropropyl)dimethoxymethylsilane 97%
(3,3,3-Trifluoropropyl)dimethoxymethylsilane
Purity: 97%
CAS No.: 358-67-8
Molecular Formula: C6H13F3O2Si
Formula Weight: 202.25
Flash Point: 58°(136°F)
Refractive Index: 1.3576
Boiling Point: 85°/150mm
Beilstein Reference: 1753237
UN No.: 1993
MDL No.: MFCD00053175
Formula:
C₆H₁₃F₃O₂Si MW: 202.25 g/mol Boiling Pt: 85 °C/150 mm Density: 1.100 Flash Pt: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00053175 CAS Number: 358-67-8 EINECS: 206-619-4 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...