Methyl-3,5-difluorobenzoate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L17171-06
L17171-14
CAAAAL17171-06EA
106.02
CAD
CAAAAL17171-06
CAAAAL17171-14
Methyl-3,5-difluorobenzoate 98%
Methyl 3,5-difluorobenzoate
Purity: 98%
CAS No.: 216393-55-4
Molecular Formula: C8H6F2O2
Formula Weight: 172.13
Refractive Index: 1.4740
Formula:
C₈H₆F₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD01318160 CAS Number: 216393-55-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...