Diethyl (bromodifluoromethyl)phosphonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16715-03
L16715-06
L16715-14
CAAAAL16715-03EA
50.4
CAD
CAAAAL16715-03
CAAAAL16715-06
CAAAAL16715-14
Diethyl (bromodifluoromethyl)phosphonate 97%
Diethyl (bromodifluoromethyl)phosphonate
Purity: 97%
CAS No.: 65094-22-6
Molecular Formula: C5H10BrF2O3P
Formula Weight: 267.01
Flash Point:
Formula:
C₅H₁₀BrF₂O₃P Boiling Pt: 99…102 °C/16 mm Density: 1.503 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00069118 CAS Number: 65094-22-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...