(3,3,3-Trifluoropropyl)dichloromethylsilane 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Dichloro(methyl)(3,3,3-trifluoropropyl)silane, (3,3,3-Trifluoropropyl)methyldichlorosilane
L16669-14
L16669-22
CAAAAL16669-06EA
78.78
CAD
CAAAAL16669-06
CAAAAL16669-14
CAAAAL16669-22
(3,3,3-Trifluoropropyl)dichloromethylsilane 97%
(3,3,3-Trifluoropropyl)dichloromethylsilane
Purity: 97%
CAS No.: 675-62-7
Molecular Formula: C4H7Cl2F3Si
Formula Weight: 211.09
Flash Point: 15°(59°F)
Refractive Index: 1.3850
Boiling Point: 121-122°
Beilstein Reference: 1744964
UN No.: 2985
MDL No.: MFCD00039294
Formula:
C₄H₇Cl₂F₃Si MW: 211.09 Boiling Pt: 121…122 °C Density: 1.261 Flash Pt: 15 °C(59 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00039294 CAS Number: 675-62-7 EINECS: 211-623-4 UN: 2985 ADR: "3",II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...