Methyl-5-methylnicotinate 94%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L16371-03
L16371-06
CAAAAL16371-03EA
93.06
CAD
CAAAAL16371-03
CAAAAL16371-06
Methyl-5-methylnicotinate 94%
Methyl 5-methylnicotinate
Purity: 94%
CAS No.: 29681-45-6
Molecular Formula: C8H9NO2
Formula Weight: 151.17
Melting Point: 42-46°
Boiling Point: 90°/3mm
MDL No.: MFCD01075739
Formula:
C₈H₉NO₂ Boiling Pt: 90 °C/3 mm Melting Pt: 42…46 °C Storage Temperature: Ambient |
MDL Number:
MFCD01075739 CAS Number: 29681-45-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...