Dodecamolybdophosphoric acid hydrate Mo ≥ 47,5%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Molybdophosphoric acid hydrate, 12-Molybdophosphoric acid hydrate, Phosphomolybdic acid hydrate

L15827-14 L15827-22 L15827-36
CAAAAL15827-14EA 70.94 CAD
CAAAAL15827-14 CAAAAL15827-22 CAAAAL15827-36
Dodecamolybdophosphoric acid hydrate Mo ≥ 47,5%
Dodecamolybdophosphoric acid hydrate

CAS No.: 51429-74-4 Molecular Formula: H3PO4.12MoO3.xH2O Formula Weight: 1825.25(anhy) Melting Point: 78-90° Merck Reference: 14,7342 UN No.: 3085 MDL No.: MFCD00149913

Formula: H₃[P(Mo₃O₁₀)₄]·nH₂O
Melting Pt: 78…90 °C
Density: 3.15
Storage Temperature: Ambient
MDL Number: MFCD00149913
CAS Number: 51429-74-4
EINECS: 234-713-5
UN: 3084
ADR: 8,II
Merck Index: 14,7342

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR