4,5-Dibromo-2-furoic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 4,5-Dibromofuran-2-carboxylic acid

L15517-06 L15517-14
CAAAAL15517-03EA 82.71 CAD
CAAAAL15517-03 CAAAAL15517-06 CAAAAL15517-14
4,5-Dibromo-2-furoic acid 97%
4,5-Dibromo-2-furoic acid

Purity: 97% CAS No.: 2434-03-9 Molecular Formula: C5H2Br2O3 Formula Weight: 269.88 Melting Point: 169-172° Beilstein Reference: 129312

Formula: C₅H₂Br₂O₃
MW: 269.88 g/mol
Melting Pt: 169…172 °C
Storage Temperature: Ambient
MDL Number: MFCD00092311
CAS Number: 2434-03-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR