3-Formyl-2-thiopheneboronic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Formylthiophene-2-boronic acid, (3-Formylthiophen-2-yl)boronic acid

L15196-03 L15196-06
CAAAAL15196-03EA 298.62 CAD
CAAAAL15196-03 CAAAAL15196-06
3-Formyl-2-thiopheneboronic acid 97%
3-Formyl-2-thiopheneboronic acid

Purity: 97% CAS No.: 17303-83-2 Molecular Formula: C5H5BO3S Formula Weight: 155.97 Storage Temperature: 2°C to 8°C Melting Point: ca 180° dec. Beilstein Reference: 4387740 MDL No.: MFCD01075679

Formula: C₅H₅BO₃S
Melting Pt: ca 180 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01075679
CAS Number: 17303-83-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR