2-Formyl-3-thiopheneboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L15195-03
L15195-06
CAAAAL15195-03EA
84.21
CAD
CAAAAL15195-03
CAAAAL15195-06
2-Formyl-3-thiopheneboronic acid 97%
2-Formyl-3-thiopheneboronic acid
Purity: 97%
CAS No.: 4347-31-3
Molecular Formula: C5H5BO3S
Formula Weight: 155.97
Storage Temperature: 2°C to 8°C
Melting Point: ca 200° dec.
Beilstein Reference: 4393584
MDL No.: MFCD01075678
Formula:
C₅H₅BO₃S Melting Pt: ca 200 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD01075678 CAS Number: 4347-31-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...