Cyclopentylacetyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
L14562-06
L14562-14
CAAAAL14562-06EA
129.72
CAD
CAAAAL14562-06
CAAAAL14562-14
Cyclopentylacetyl chloride 97%
Cyclopentylacetyl chloride
Purity: 97%
CAS No.: 1122-99-2
Molecular Formula: C7H11ClO
Formula Weight: 146.62
Flash Point: 71°(160°F)
Refractive Index: 1.4625
Odor: Pungent
Boiling Point: 186°
Beilstein Reference: 1906113
UN No.: 3265
MDL No.: MFCD00236031
Formula:
C₇H₁₁ClO Boiling Pt: 186 °C Density: 1.087 Flash Pt: 71 °C(160 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00236031 CAS Number: 1122-99-2 UN: 3265 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...