1-Naphthonitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
L14498-06
L14498-14
CAAAAL14498-06EA
41.71
CAD
CAAAAL14498-06
CAAAAL14498-14
1-Naphthonitrile 95%
1-Naphthonitrile
Purity: 95%
CAS No.: 86-53-3
Molecular Formula: C11H7N
Formula Weight: 153.18
Melting Point: 33-38°
Boiling Point: 299°
UN No.: 3439
MDL No.: MFCD00003866
Formula:
C₁₁H₇N Boiling Pt: 299 °C Melting Pt: 33…38 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003866 CAS Number: 86-53-3 EINECS: 201-679-8 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...