Kerosene low odor
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
C13-14 isoparaffin, Light distillate
L14479-0F
L14479-AP
CAAAAL14479-0FEA
252.36
CAD
CAAAAL14479-0F
CAAAAL14479-AP
Kerosene low odor
Kerosene - CAS 64742-47-8
CAS No.: 64742-47-8
Flash Point: 76°(169°F)
Refractive Index: 1.4440
Boiling Point: 200-250°
Merck Reference: 14,5294
UN No.: 1223
MDL No.: MFCD00135561
Boiling Pt:
200…250 °C Density: 0.800 Flash Pt: 76 °C(169 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00135561 CAS Number: 64742-47-8 EINECS: 265-149-8 Merck Index: 14,5294 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...