Trimellitic anhydride chloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
1,3-Dioxo-1,3-dihydroisobenzofuran-5-carbonyl chloride, 1,2,4-Benzenetricarboxylic acid 1,2-anhydride 4-chloride
L14382-14
L14382-22
CAAAAL14382-14EA
71.89
CAD
CAAAAL14382-14
CAAAAL14382-22
Trimellitic anhydride chloride 98%
Trimellitic anhydride chloride
Purity: 98%
CAS No.: 1204-28-0
Molecular Formula: C9H3ClO4
Formula Weight: 210.57
Storage Temperature: 2°C to 8°C
Melting Point: 67-69°
Beilstein Reference: 610823
UN No.: 3261
MDL No.: MFCD00005924
Formula:
C₉H₃ClO₄ MW: 210.57 g/mol Melting Pt: 67…69 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005924 CAS Number: 1204-28-0 EINECS: 214-874-8 UN: 3261 ADR: "8",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...